Nickel, gauze woven from wire 20 mesh, Ø 0.18 mm (0.007 in)
Supplier: Thermo Fisher Scientific
Danger
39704.0S 39704.WO
39704.0SEA 311 EUR
39704.0S 39704.WO
Nickel, gauze woven from wire 20 mesh, Ø 0.18 mm (0.007 in)
Nickel
CAS No.: 7440-02-0 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582