Nickel 99,8 Metals basis, powder_spherical -100+325 mesh
Supplier: Thermo Fisher Scientific
Danger
42733.30 42733.A1
42733.30EA 119 EUR
42733.30 42733.A1
Nickel 99,8 Metals basis, powder_spherical -100+325 mesh
Nickel
Purity: 99.8% CAS No.: 7440-02-0 Appearance: Gray Odour: Odourless UN No.: 3089 MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 II
Merck Index: 12,06582