Nickel ≥99.98% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific
Danger
43131.H4 43131.G9 43131.G5
43131.H4EA 572 EUR
43131.H4 43131.G9 43131.G5
Nickel ≥99.98% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Nickel
Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈0.44g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582