Nickel 99,5 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Danger
44821.FI 44821.GH 44821.HP
44821.FIEA 30.1 EUR
44821.FI 44821.GH 44821.HP
Nickel 99,5 Metals basis, foil, Thickness: 0.1 mm (0.004 in)
Nickel
Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈2.2g/50x50mm Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582