Nickel ≥99.5% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Danger
44825.FI 44825.GH 44825.HP
44825.FIEA 66.1 EUR
44825.FI 44825.GH 44825.HP
Nickel ≥99.5% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Nickel
Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈44.5g/50x50mm Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582