Nickel, gauze_wire 60 mesh, Ø 0.18 mm (0.007 in)
Supplier: Thermo Fisher Scientific
Danger
45099.HB 45099.FL 45099.GT
45099.HBEA 427 EUR
45099.HB 45099.FL 45099.GT
Nickel, gauze_wire 60 mesh, Ø 0.18 mm (0.007 in)
Nickel
CAS No.: 7440-02-0 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582