Nickel, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
267074-14G
267074-42G
SIAL267074-14GEA
163
EUR
SIAL267074-14G
SIAL267074-42G
Nickel, Sigma-Aldrich®
Nickel
Formula:
Ni Boiling Pt: 3007 °C (1013 hPa) Melting Pt: 1453 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |