Nickel, Sigma-Aldrich®
Supplier: Merck
Danger
MQ100
357588-2.2G 357588-8.8G
SIAL357588-2.2GEA 277 EUR
SIAL357588-2.2G SIAL357588-8.8G
Nickel, Sigma-Aldrich®
Nickel
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582