Palladium ≥99.9% (metals basis), wire, Ø 0.5 mm
Supplier: Thermo Fisher Scientific
10279.BW 10279.CB 10279.CD
10279.BWEA 448 EUR
10279.BW 10279.CB 10279.CD
Palladium ≥99.9% (metals basis), wire, Ø 0.5 mm
Palladium
Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈2.36g/m Odour: Odourless MDL No.: MFCD00011167
Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121