Palladium ≥99.99% (metals basis), wire, Premion®, Ø 0.1 mm
Supplier: Thermo Fisher Scientific
10963.G5 10963.G9
10963.G5EA 173 EUR
10963.G5 10963.G9
Palladium ≥99.99% (metals basis), wire, Premion®, Ø 0.1 mm
Palladium
Purity: 99.99% CAS No.: 7440-05-3 Physical Form: ≈0.09g/m Odour: Odourless MDL No.: MFCD00011167
Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121