Platinum 99,9 Metals basis, foil, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
00261.FF 00261.FI
00261.FFEA 545 EUR
00261.FF 00261.FI
Platinum 99,9 Metals basis, foil, Thickness: 0.127 mm (0.005 in)
Platinum
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈1.7g/25x25mm Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689