Platinum ≥99.9% (metals basis), gauze woven from wire 100 mesh, Ø 0.0762 mm (0.003 in)
Supplier: Thermo Fisher Scientific
10282.FF 10282.FI 10282.GH
10282.FFEA 485 EUR
10282.FF 10282.FI 10282.GH
Platinum ≥99.9% (metals basis), gauze woven from wire 100 mesh, Ø 0.0762 mm (0.003 in)
Platinum
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.53g/25x25mm, Wire Cloth Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689