Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
11509.FF 11509.FI 11509.GH
11509.FFEA 506 EUR
11509.FF 11509.FI 11509.GH
Platinum ≥99.99% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
Platinum
Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈1.34g/25x25mm Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689