Platinum 99,995 Metals basis, foil, Premion®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
12058.FF 12058.FI
12058.FFEA 1070 EUR
12058.FF 12058.FI
Platinum 99,995 Metals basis, foil, Premion®, Thickness: 0.25 mm (0.01 in)
Platinum
Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈3.35g/25x25mm Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689