Platinum, black, HiSPEC® 1000
Supplier: Thermo Fisher Scientific
12755.03
12755.06
12755.14
12755.01
12755.03EA
0
EUR
12755.03
12755.06
12755.14
12755.01
Platinum, black, HiSPEC® 1000
Platinum
CAS No.: 7440-06-4
Physical Form: S.A. nominally 27m2/g
Odour: Odourless
MDL No.: MFCD00011179
Formula:
Pt Boiling Pt: 4530 °C (1013 hPa) Melting Pt: 1773 °C Density: 21.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 UN: 3089 ADR: 4.1,II |