Platinum, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
349356-2.4G 349356-600MG
SIAL349356-2.4GEA 1680 EUR
SIAL349356-2.4G SIAL349356-600MG
Platinum, Sigma-Aldrich®
Platinum
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689