Platinum, Sigma-Aldrich®
Supplier: Merck
Synonyms: Platinium
M-Clarity™ quality level = MQ100
349364-1.4G 349364-350MG
SIAL349364-1.4GEA 1230 EUR
SIAL349364-1.4G SIAL349364-350MG
Platinum, Sigma-Aldrich®
Platinum
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689