Platinum, Sigma-Aldrich®

Supplier: Merck

Synonyms: Platinium

M-Clarity™ quality level = MQ100

349402-1.5G 349402-250MG
SIAL349402-1.5GEA 471 EUR
SIAL349402-1.5G SIAL349402-250MG
Platinum, Sigma-Aldrich®
Platinum
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR