Platinum, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
357375-100MG
357375-500MG
SIAL357375-100MGEA
110
EUR
SIAL357375-100MG
SIAL357375-500MG
Platinum, Sigma-Aldrich®
Platinum
Formula:
Pt Boiling Pt: 3827 °C (1013 hPa) Melting Pt: 1769 °C Density: 21.447 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...