Platinum, powder, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
327476-500MG 327476-5G
SIAL327476-500MGEA 231 EUR
SIAL327476-500MG SIAL327476-5G
Platinum, powder, Sigma-Aldrich®
Platinum
Formula: Pt
Boiling Pt: 4530 °C (1013 hPa)
Melting Pt: 1773 °C
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II