Rhodium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific
11575.BP 11575.BS
11575.BPEA 562 EUR
11575.BP 11575.BS
Rhodium ≥99.8% (metals basis), wire, Ø 1.0 mm (0.04 in)
Rhodium
Purity: 99.8% CAS No.: 7440-16-6 Physical Form: ≈9.74g/m MDL No.: MFCD00011201
Formula: Rh
MW: 102.9 g/mol
Boiling Pt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960 °C
Density: 12.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353