Ruthenium ≥99.95% (metals basis), sponge -20 mesh
Supplier: Thermo Scientific
Warning
12062.04 12062.09 12062.18
12062.04EA 267 EUR
12062.04 12062.09 12062.18
Ruthenium ≥99.95% (metals basis), sponge -20 mesh
Ruthenium
Purity: 99.95% CAS No.: 7440-18-8 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011207
Formula: Ru
MW: 101.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II
Merck Index: 12,08452