Ruthenium 99.9% (trace metals basis), powder -200 mesh
Supplier: Thermo Scientific
Warning
198100010 198100050 198100250
ACRO198100010EA 143 EUR
ACRO198100010 ACRO198100050 ACRO198100250
Ruthenium 99.9% (trace metals basis), powder -200 mesh
Ruthenium
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12,45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452
Order Now
Specification Test Results
Appearance Grey Powder
Total trace metal impurities ≤0.1 %