Ruthenium ≤100 ppm ≥99.98% (metals basis excluding Ca), powder, Premion® -22 mesh
Supplier: Thermo Scientific
Warning
10667.06 10667.14
10667.06EA 667 EUR
10667.06 10667.14
Ruthenium ≤100 ppm ≥99.98% (metals basis excluding Ca), powder, Premion® -22 mesh
Ruthenium
Purity: 99.98% CAS No.: 7440-18-8 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011207
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452