Ruthenium ≥99.9% (metals basis), powder -325 mesh
Supplier: Thermo Scientific
Warning
13994.04 13994.09 13994.18
13994.04EA 267 EUR
13994.04 13994.09 13994.18
Ruthenium ≥99.9% (metals basis), powder -325 mesh
Ruthenium
Purity: 99.9% CAS No.: 7440-18-8 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011207
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II
Merck Index: 12,08452