Ruthenium ≥99.99% (metals basis excluding Ca), powder, Premion® -200 mesh (max. 0.01% Ca)
Supplier: Thermo Scientific
Warning
44879.06 44879.03 44879.14
44879.06EA 667 EUR
44879.06 44879.03 44879.14
Ruthenium ≥99.99% (metals basis excluding Ca), powder, Premion® -200 mesh (max. 0.01% Ca)
Ruthenium
Purity: 99.99+% CAS No.: 7440-18-8 Appearance: Grey Odour: Odourless UN No.: 3089 MDL No.: MFCD00011207
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452