Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼2 mm), indicator_with
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
32256.30 32256.A1 32256.A7
32256.30EA 78.3 EUR
32256.30 32256.A1 32256.A7
Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12 (∼2 mm), indicator_with
Molecular sieve A4 (0.4 nm, 4 Å)
Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0
UN: 3077
ADR: 9 III