DL-Isoleucine, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-DL-Ile-OH
M-Clarity™ quality level = MQ200
298689-10G 298689-50G
SIAL298689-10GEA 56.3 EUR
SIAL298689-10G SIAL298689-50G
DL-Isoleucine, Sigma-Aldrich®
DL-Isoleucine
Formula: C₆H₁₃NO₂
Melting Pt: 275 °C
Density: 1.255 g/cm³ (–123.15 °C)
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8
Merck Index: 13,05198