1,2-Dibromoethylene, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetylene dibromide

D40809-25G D40809-5G
SIALD40809-25GEA 170 EUR
SIALD40809-25G SIALD40809-5G
1,2-Dibromoethylene, Sigma-Aldrich®
1,2-Dibromoethylene
Formula: C₂H₂Br₂
MW: 185.85 g/mol
Boiling Pt: 111…112 °C (1013 hPa)
Melting Pt: –53 °C
Density: 2.268 g/cm³ (0 °C)
MDL Number: MFCD00000184
CAS Number: 540-49-8
EINECS: 208-747-6
UN: 2810
ADR: 6.1 III
Merck Index: 12,00092

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR