trans-2-Decenal (synthetic) ≥95% FCC, FG (Halal, Kosher), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: trans-2-Decenal, (E)-2-Decenal, (E)-Dec-2-enal
M-Clarity™ quality level = MQ300
W236608-1KG-K W236608-200G-K
SIALW236608-1KG-KEA 771 EUR
SIALW236608-1KG-K SIALW236608-200G-K
trans-2-Decenal (synthetic) ≥95% FCC, FG (Halal, Kosher), Sigma-Aldrich®
trans-2-Decenal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 78…80 °C (3 mmHg) (lit.)
Density: 0.841 g/mL (25 °C) (lit.)
Flash Pt: 96 °C
MDL Number: MFCD00014679
CAS Number: 3913-81-3
EINECS: 223-472-1
UN: 3082
ADR: 9 III