Crotyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Buten-1-ol, 1-Hydroxy-2-butene

MQ200

C86006-100ML C86006-25ML
SIALC86006-100MLEA 160 EUR
SIALC86006-100ML SIALC86006-25ML
Crotyl alcohol, Sigma-Aldrich®
Crotyl alcohol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 120
Melting Pt: < –30 °C
Density: 0.8726 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002923
CAS Number: 6117-91-5
EINECS: 228-086-7
UN: 1987
ADR: 3 III
Merck Index: 13,02629

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR