2-Pentene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
143774-100G 143774-25G
SIAL143774-100GEA 314 EUR
SIAL143774-100G SIAL143774-25G
2-Pentene, Sigma-Aldrich®
2-Pentene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 37
Melting Pt: –140 °C
Density: 0.658 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009384
CAS Number: 109-68-2
EINECS: 203-695-0
UN: 3295
ADR: 3 II
Merck Index: 12,07263