Isoeugenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Propenylguaiacol, 4-Hydroxy-3-methoxy-1-propenylbenzene, 2-Methoxy-4-(prop-1-en-1-yl)phenol
M-Clarity™ quality level = MQ300
W246808-10KG-K W246808-1KG-K
SIALW246808-10KG-KEA 1230 EUR
SIALW246808-10KG-K SIALW246808-1KG-K
Isoeugenol, Sigma-Aldrich®
Isoeugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 266
Melting Pt: –10 °C
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 112
Storage Temperature: Ambient
MDL Number: MFCD00009285
CAS Number: 97-54-1
EINECS: 202-590-7
Merck Index: 13,05189