trans-1,2-Diphenylpropene ≥98%
Supplier: Thermo Fisher Scientific
Synonyms: (E)-α-Methylstilbene, (E)-Prop-1-ene-1,2-diyldibenzene
A15351.06 A15351.14
A15351.06EA 82.8 EUR
A15351.06 A15351.14
trans-1,2-Diphenylpropene ≥98%
trans-1,2-Diphenylpropene
Purity: 98% CAS No.: 833-81-8 Molecular Formula: C15H14 Formula Weight: 194.28 Melting Point: 79-82° Beilstein Reference: 1906425
Formula: C₁₅H₁₄
MW: 194.28 g/mol
Melting Pt: 79…82 °C
Density: 0.986
Storage Temperature: Ambient
MDL Number: MFCD00026343
CAS Number: 833-81-8
EINECS: 212-300-0