1,5-Cyclooctadiene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cycloocta-1,5-diene
M-Clarity™ quality level = MQ100
246050-100ML 246050-800ML
SIAL246050-100MLEA 58.5 EUR
SIAL246050-100ML SIAL246050-800ML
1,5-Cyclooctadiene, Sigma-Aldrich®
1,5-Cyclooctadiene
Formula: C₈H₁₂
MW: 108.18 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –69 °C
Density: 0.8811 g/cm³ (20 °C)
MDL Number: MFCD00001752
CAS Number: 111-78-4
EINECS: 203-907-1
UN: 2520
ADR: 3,III