1,5-Cyclooctadiene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Cycloocta-1,5-diene
M-Clarity™ quality level = MQ100
C109207-100ML
C109207-1L
C109207-4L
C109207-500ML
SIALC109207-100MLEA
46.8
EUR
SIALC109207-100ML
SIALC109207-1L
SIALC109207-4L
SIALC109207-500ML
1,5-Cyclooctadiene, Sigma-Aldrich®
1,5-Cyclooctadiene
Formula:
C₈H₁₂ MW: 108.18 g/mol Boiling Pt: 151 °C (1013 hPa) Melting Pt: –69 °C Density: 0.8811 g/cm³ (20 °C) |
MDL Number:
MFCD00001752 CAS Number: 111-78-4 EINECS: 203-907-1 UN: 2520 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...