Decahydronaphthalene (mixture of cis and trans isomers), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene
M-Clarity™ quality level = MQ100
294772-100ML 294772-1L 294772-2L
SIAL294772-100MLEA 112 EUR
SIAL294772-100ML SIAL294772-1L SIAL294772-2L
Decahydronaphthalene (mixture of cis and trans isomers), Sigma-Aldrich®
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
Boiling Pt: 189…191 °C (1013 hPa)
Melting Pt: ∼–40 °C
Density: 0.8696 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3,III
Merck Index: 13,02866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM