1,2-Diaminocyclohexane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Diaminocyclohexane, 1,2-Cyclohexanediamine
M-Clarity™ quality level = MQ100
132551-1L 132551-250ML 132551-50ML
SIAL132551-1LEA 229 EUR
SIAL132551-1L SIAL132551-250ML SIAL132551-50ML
1,2-Diaminocyclohexane, Sigma-Aldrich®
1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0.93 g/cm³ (20 °C)
MDL Number: MFCD00001491
CAS Number: 694-83-7
EINECS: 211-776-7