1,4-Cyclohexanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cyclohexane-1,4-diol
M-Clarity™ quality level = MQ100
C101206-100G C101206-25G
SIALC101206-100GEA 58.8 EUR
SIALC101206-100G SIALC101206-25G
1,4-Cyclohexanediol, Sigma-Aldrich®
1,4-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Density: 1.17 g/cm³ (21.85 °C)
MDL Number: MFCD00001448
CAS Number: 556-48-9
EINECS: 209-126-2