2-Methylcyclohexanol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Methylcyclohexan-1-ol
M-Clarity™ quality level = MQ100
153087-1L
153087-250ML
SIAL153087-1LEA
285
EUR
SIAL153087-1L
SIAL153087-250ML
2-Methylcyclohexanol, Sigma-Aldrich®
2-Methylcyclohexanol
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 163 Melting Pt: –38 °C Density: 0.921 g/cm³ (20 °C) Flash Pt: 58 Storage Temperature: Ambient |
MDL Number:
MFCD00001443 CAS Number: 583-59-5 EINECS: 209-512-0 UN: 2617 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...