2,5-Dimethoxytetrahydrofuran, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
D137103-100G
D137103-500G
SIALD137103-100GEA
63
EUR
SIALD137103-100G
SIALD137103-500G
2,5-Dimethoxytetrahydrofuran, Sigma-Aldrich®
2,5-Dimethoxytetrahydrofuran
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –45 °C Density: 1.021 g/cm³ (20 °C) Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005359 CAS Number: 696-59-3 EINECS: 211-797-1 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...