Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)
Supplier: Thermo Scientific
Synonyms: Linde 13X
33550.A1 33550.A7
33550.A1EA 121 EUR
33550.A1 33550.A7
Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)
Molecular sieve 13X (1.0 nm, 10 Å)
Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6