Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 1-2mm (0.04-0.08in)
Supplier: Thermo Scientific
Synonyms: Linde 13X
87954.A1 87954.A7
87954.A1EA 137 EUR
87954.A1 87954.A7
Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 1-2mm (0.04-0.08in)
Molecular sieve 13X (1.0 nm, 10 Å)
Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6