Molecular sieve 13X (1.0 nm, 10 Å) 3-5 mm (0.12-0.20 in)
Supplier: Thermo Scientific
Synonyms: Linde 13X
L06085.30 L06085.0B L06085.0I
L06085.30EA 47.2 EUR
L06085.30 L06085.0B L06085.0I
Molecular sieve 13X (1.0 nm, 10 Å) 3-5 mm (0.12-0.20 in)
Molecular sieve 13X (1.0 nm, 10 Å)
CAS No.: 63231-69-6 MDL No.: MFCD00131613
Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6