Molecular sieve 3A (0.3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)
Supplier: Thermo Scientific
Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
L05359.30 L05359.0B
L05359.30EA 57.2 EUR
L05359.30 L05359.0B
Molecular sieve 3A (0.3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve 3A (0.3 nm, 3 Å)
CAS No.: 308080-99-1 MDL No.: MFCD00131613
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1