Paraffin, liquid, TECHNICAL
Supplier: VWR Chemicals
Danger
24677.294EA 78.9 EUR
24677.294 24677.363
Paraffin, liquid, TECHNICAL
Paraffin
Boiling Pt: 300-500 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Flash Pt: 215 °C (closed cup)
Storage Temperature: Ambient
CAS Number: 8042-47-5
EINECS: 232-384-2
Merck Index: 13,07093
Order Now
Specification Test Results
Identification Passes test