Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Ø 3 - 4mm (0.12-0.16in)
Supplier: Thermo Scientific
Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
88120.A1 88120.A7
88120.A1EA 144 EUR
88120.A1 88120.A7
Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Ø 3 - 4mm (0.12-0.16in)
Molecular sieve A4 (0.4 nm, 4 Å)
Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0