Molecular sieve A4 (0.4 nm, 4 Å) 3 - 5 mm (0.12-0.20 in)
Supplier: Thermo Fisher Scientific
Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
L05466.30 L05466.0B L05466.0I
L05466.30EA 49.9 EUR
L05466.30 L05466.0B L05466.0I
Molecular sieve A4 (0.4 nm, 4 Å) 3 - 5 mm (0.12-0.20 in)
Molecular sieve A4 (0.4 nm, 4 Å)
CAS No.: 70955-01-0 MDL No.: MFCD00131613
Density: 1.1…2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0