Petroleum spirit, 40…60 °C
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Petroleum ether, Petroleum benzine
L14649.AU L14649.0F
L14649.AUEA 38.7 EUR
L14649.AU L14649.0F
Petroleum spirit, 40…60 °C
Petroleum spirit (8032-32-4)
CAS No.: 8032-32-4 Flash Point: -40°(-40°F) Odour: Petroleum-like Boiling Point: 40-60° UN No.: 1268 MDL No.: MFCD00081849
Boiling Pt: 40…60 °C
Density: 0.660
Flash Pt: –40 °C (–40 °F)
Storage Temperature: Ambient
CAS Number: 8032-32-4
EINECS: 232-453-7
UN: 1268
ADR: 3 II