Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Petroleum ether, Petroleum benzine
L13734.AU L13734.0F
L13734.AUEA 33.4 EUR
L13734.AU L13734.0F
Petroleum spirit, 60…80 °C
Petroleum spirit (8032-32-4)
CAS No.: 8032-32-4 Flash Point: -10°(14°F) Odour: Petroleum-like Boiling Point: 60-80° Merck Reference: 14,5489 UN No.: 1268 MDL No.: MFCD00081849
Boiling Pt: 60…80 °C
Density: 0.672
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
CAS Number: 8032-32-4
EINECS: 232-453-7
UN: 1268
ADR: 3 II
Merck Index: 14,5489